APO455837446 Apollo Scientific Ltd

Ethyl 3-chloro-4-fluorobenzoate

  • MDL number MFCD06204341
  • Cas Number : 137521-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-40
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-chloro-4-fluorobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-chloro-4-fluorobenzoate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart