APO455837447 Apollo Scientific Ltd

(2R)-2-Hydroxy-2-(trifluoromethyl)propanoic acid

  • MDL number MFCD03095400
  • Cas Number : 44864-47-3
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Synonym (2R)-2-Hydroxy-2-methyl-3,3,3-trifluoropropanoic acid, (2R)-2-Hydroxy-2-methyl-3,3,3-trifluoropropionic acid
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 189
Product Link https://store.apolloscientific.co.uk/product/2r-2-hydroxy-2-trifluoromethylpropanoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2r-2-hydroxy-2-trifluoromethylpropanoic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart