APO455837452 Apollo Scientific Ltd

1H-Perfluoroheptane

  • MDL number MFCD00042108
  • Cas Number : 375-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-Pentadecafluoroheptane, 1H-Pentadecafluoroheptane
Purity 97
Density 1.725
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 96-97
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4410

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4410
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart