APO455837457 Apollo Scientific Ltd

Perfluorohept-1-ene

  • MDL number MFCD00013577
  • Cas Number : 355-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1,2,3,3,4,4,5,5,6,6,7,7,7-Tetradecafluorohept-1-ene
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 80-82
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4424

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4424
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart