APO455837458 Apollo Scientific Ltd

N-Ethyl-4-(trifluoromethoxy)aniline

  • MDL number MFCD00203992
  • Cas Number : 175278-20-3
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N[4-(Trifluoromethoxy)]ethylamine
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42-44/0.2mm
Product Link https://store.apolloscientific.co.uk/product/n-ethyl-4-trifluoromethoxyaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/n-ethyl-4-trifluoromethoxyaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart