APO455837459 Apollo Scientific Ltd

6-Fluoro-3-(piperidin-4-yl)-1,2-benzisoxazole hydrochloride

  • MDL number MFCD06658529
  • Cas Number : 84163-13-3
  • Molecular Weight : 256.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 293-295
Synonym 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidine hydrochloride, 6-Fluoro-3-(piperidin-4-yl)benzo[d]isoxazole hydrochloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-fluoro-3-piperidin-4-yl-12-benzisoxazole-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-fluoro-3-piperidin-4-yl-12-benzisoxazole-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart