APO455837461 Apollo Scientific Ltd

5-Chloro-3-fluoropyridine-2-carbonitrile

  • MDL number MFCD07375055
  • Cas Number : 207994-11-4
  • Molecular Weight : 156.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-73
Synonym 5-Chloro-2-cyano-3-fluoropyridine
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-3-fluoropyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-3-fluoropyridine-2-carbonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart