APO455837466 Apollo Scientific Ltd

Perfluorohexadecane

  • MDL number MFCD00042085
  • Cas Number : 355-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 239-240
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4432

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4432
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart