APO455837471 Apollo Scientific Ltd

5-Chloro-3-fluoropyridine-2-carboxylic acid

  • MDL number MFCD12827555
  • Cas Number : 207994-08-9
  • Molecular Weight : 175.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-174
Synonym 5-Chloro-3-fluoropicolinic acid, 2-Carboxy-5-chloro-3-fluoropyridine
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-3-fluoropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-3-fluoropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart