APO455837475 Apollo Scientific Ltd

5-Bromo-2-nitro-4-(trifluoromethoxy)aniline

  • MDL number MFCD12756436
  • Cas Number : 755030-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-96
Synonym 2-Amino-4-bromo-5-(trifluoromethoxy)nitrobenzene
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46261

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46261
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart