APO455837481 Apollo Scientific Ltd

3-Perfluorohexyl-2-iodopropan-1-ol

  • MDL number MFCD00236070
  • Cas Number : 38550-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49
Purity 97
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 90/1.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4454

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4454
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart