APO455837483 Apollo Scientific Ltd

5-Methoxy-2-nitrobenzotrifluoride

  • MDL number MFCD00053611
  • Cas Number : 344-39-8
  • Molecular Weight : 221.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48
Synonym 4-Nitro-3-(trifluoromethyl)anisole
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methoxy-2-nitrobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methoxy-2-nitrobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart