APO455837486 Apollo Scientific Ltd

Perfluoro(methylcyclopentane)

  • MDL number MFCD00066611
  • Cas Number : 1805-22-7
  • Molecular Weight : 300.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -90
Purity 90
Density 1.68
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 47-53
Product Link https://store.apolloscientific.co.uk/product/perfluoromethylcyclopentane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/perfluoromethylcyclopentane
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart