APO455837489 Apollo Scientific Ltd

1H,1H,2H,2H-Hexadecafluoro-9-(trifluoromethyl)decyl iodide

  • MDL number MFCD00153247
  • Cas Number : 40678-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H,1H,2H,2H-Perfluoro-9-methyldecyl iodide, 1,1,1,2,3,3,4,4,5,5,6,6,7,7,8,8-Hexadecafluoro-10-iodo-2-(trifluoromethyl)decane
Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4485

Safety Information

signalword Warning
pcodes P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4485
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart