APO455837491 Apollo Scientific Ltd

Perfluoro(N-methylmorpholine)

  • MDL number MFCD00042442
  • Cas Number : 382-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,3,3,5,5,6,6-octafluoro-4-(trifluoromethyl)morpholine
Purity 98
Density 1.7
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 50
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4498

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4498
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart