APO455837495 Apollo Scientific Ltd

4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluorononane-1,2-diol

  • MDL number MFCD00042262
  • Cas Number : 107650-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Synonym 1H,1H,2H,3H,3H-Perfluorononane-1,2-diol, 3-(Perfluorohex-1-yl)propane-1,2-diol
Purity 97
Flash Point >150
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 120-130
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4516

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4516
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart