APO455837502 Apollo Scientific Ltd

5-(Bromomethyl)-2-fluorobenzonitrile

  • MDL number MFCD03094341
  • Cas Number : 180302-35-6
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-4-fluorobenzyl bromide, alpha-Bromo-6-fluoro-m-tolunitrile
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromomethyl-2-fluorobenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromomethyl-2-fluorobenzonitrile
pictograms GHS05;GHS07
hstatements H302;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart