APO455837506 Apollo Scientific Ltd

3-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine

  • MDL number MFCD11111688
  • Cas Number : 503172-42-7
  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-trifluoromethylimidazo12-apyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-trifluoromethylimidazo12-apyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart