APO455837516 Apollo Scientific Ltd

(Perfluoro-n-octyl)ethane

  • MDL number MFCD00236711
  • Cas Number : 77117-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H,1H,1H,2H,2H-Perfluorodecane
Density 1.654
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 77/50mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4569

Safety Information

signalword Warning
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4569
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart