APO455837521 Apollo Scientific Ltd

4-Bromo-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD07787468
  • Cas Number : 320-31-0
  • Molecular Weight : 269.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-121
Synonym 5-Bromo-2-carboxybenzotrifluoride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart