APO455837526 Apollo Scientific Ltd

3-Amino-2-fluorobenzoic acid

  • MDL number MFCD06797827
  • Cas Number : 914223-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45888

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=45888
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart