APO455837527 Apollo Scientific Ltd

3-Bromo-5-methylbenzotrifluoride

  • MDL number MFCD01631585
  • Cas Number : 86845-28-5
  • Molecular Weight : 239.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-methyl-5-(trifluoromethyl)benzene, 5-Bromo-alpha,alpha,alpha-trifluoro-m-xylene
Purity 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methylbenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methylbenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart