APO455837530 Apollo Scientific Ltd

2,6-Difluorobenzotrifluoride

  • MDL number MFCD03412213
  • Cas Number : 64248-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,3-Difluoro-2-(trifluoromethyl)benzene, alpha,alpha,alpha,2,6-Pentafluorotoluene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 106-107
Product Link https://store.apolloscientific.co.uk/product/26-difluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluorobenzotrifluoride
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart