APO455837535 Apollo Scientific Ltd

Perfluorosebaconitrile

  • MDL number MFCD00042366
  • Cas Number : 2342-09-8
  • Molecular Weight : 452.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Hexadecafluorodecane-1,10-dinitrile, Hexadecafluorooctane-1,10-dicarbonitrile
Purity 97
Density 1.67
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 147-148
Product Link https://store.apolloscientific.co.uk/product/perfluorosebaconitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/perfluorosebaconitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart