APO455837536 Apollo Scientific Ltd

Diethyl 4,4-difluoroheptane-1,7-dioate

  • MDL number MFCD08146634
  • Cas Number : 22515-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Density 1.1
Flash Point 105
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 108/3mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45946

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=45946
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart