APO455837539 Apollo Scientific Ltd

4-(Methylsulphonyl)-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD12196930
  • Cas Number : 904311-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-172
Synonym 5-Cyano-2-(methylsulphonyl)benzotrifluoride, 4-Cyano-2-(trifluoromethyl)phenyl methyl sulphone
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46041

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46041
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart