APO455837544 Apollo Scientific Ltd

Perfluoro(1,3,5-trimethylcyclohexane)

  • MDL number MFCD00066614
  • Cas Number : 374-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -68
Density 1.888
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 124-128
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4625

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4625
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart