APO455837545 Apollo Scientific Ltd

Perfluoro(2,5,8-trimethyl-3,6,9-trioxadodecanoyl)fluoride

  • MDL number MFCD00084743
  • Cas Number : 27639-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 158-161
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4629

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=4629
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart