APO455837554 Apollo Scientific Ltd

4-Fluorobenzo[b]furan-3(2H)-one

  • MDL number MFCD12026471
  • Cas Number : 911826-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Dihydro-4-fluorobenzo[b]furan-3-one, 4-Fluorocoumaran-3-one, 4-Fluoro-2,3-dihydro-3-oxo-1-benzofuran
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45457

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=45457
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart