APO455837556 Apollo Scientific Ltd

5-Fluorobenzo[b]furan-3(2H)-one

  • MDL number MFCD08544409
  • Cas Number : 60770-49-2
  • Molecular Weight : 152.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 102-106
Iupac Name 5-fluoro-1-benzofuran-3(2H)-one
Inchi Code 1S/C8H5FO2/c9-5-1-2-8-6(3-5)7(10)4-11-8/h1-4,10H
Inchi Key AVEDWWOETFRMOC-UHFFFAOYSA-N

Description

Synonym 5-Fluoro-1-benzofuran-3(2H)-one, 2,3-Dihydro-5-fluoro-3-oxobenzo[b]furan
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 102-106
Product Link https://store.apolloscientific.co.uk/product/5-fluorobenzobfuran-32h-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluorobenzobfuran-32h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart