APO455837559 Apollo Scientific Ltd

6-Fluorobenzo[b]furan-3(2H)-one

  • MDL number MFCD09743984
  • Cas Number : 351528-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Fluoro-1-benzofuran-3(2H)-one, 2,3-Dihydro-6-fluoro-3-oxobenzo[b]furan
Purity 98
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45461

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=45461
Availability details for
Quantity
  • Availability
  • Add to Cart