APO455837560 Apollo Scientific Ltd

6-Methyl-1,1,1-trifluoroheptane-2,4-dione

  • MDL number MFCD00045107
  • Cas Number : 461-92-7
  • Molecular Weight : 196.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 57
Iupac Name 1,1,1-trifluoro-6-methyl-2,4-heptanedione
density 1.13
Inchi Code 1S/C8H11F3O2/c1-5(2)3-6(12)4-7(13)8(9,10)11/h5H,3-4H2,1-2H3
Inchi Key XWBVTGFPQXBNOZ-UHFFFAOYSA-N

Description

Melting Point 57
Synonym 4-Methyl-1-(trifluoroacetyl)pentan-2-one, 2,4-Dioxo-6-methyl-1,1,1-trifluoroheptane
Density 1.13
Flash Point 78/64mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-methyl-111-trifluoroheptane-24-dione

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-methyl-111-trifluoroheptane-24-dione
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart