APO455837566 Apollo Scientific Ltd

3'-Bromo-4'-(trifluoromethoxy)acetanilide

  • MDL number MFCD13176598
  • Cas Number : 755030-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-87
Synonym 4-Acetamido-2-bromo-alpha,alpha,alpha-trifluoroanisole, N-[3-Bromo-4-(trifluoromethoxy)phenyl]acetamide
Purity 99
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46344

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46344
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart