APO455837570 Apollo Scientific Ltd

2,4-Difluorostyrene

  • MDL number MFCD00142824
  • Cas Number : 399-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Difluoro-1-vinylbenzene, 2,4-Difluoro-1-ethenylbenzene
Storage temp. Store at 2-8
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 60-62/10mm
Product Link https://store.apolloscientific.co.uk/product/24-difluorostyrene

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluorostyrene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart