APO455837579 Apollo Scientific Ltd

2-Fluoro-3-(trifluoromethoxy)benzyl bromide

  • MDL number MFCD12026474
  • Cas Number : 1159512-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-2-fluoro-3-(trifluoromethoxy)benzene, 3-(Bromomethyl)-2-fluorophenyl trifluoromethyl ether, alpha-Bromo-2-fluoro-3-(trifluoromethoxy)toluene
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45527

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45527
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart