APO455837582 Apollo Scientific Ltd

5-Chloro-1,6-dihydro-6-oxo-1-[4-(trifluoromethyl)benzyl]pyridine-3-carboxylic acid

  • MDL number MFCD04038241
  • Cas Number : 886761-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Synonym 5-Chloro-1,6-dihydro-6-oxo-1-[4-(trifluoromethyl)benzyl]nicotinic acid, 4-[5-Carboxy-3-chloro-2-oxopyridin-1(2H)-yl)methyl]benzotrifluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2786

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2786
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart