APO455837584 Apollo Scientific Ltd

5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde

  • MDL number MFCD01935951
  • Cas Number : 321848-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2790

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2790
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart