APO455837595 Apollo Scientific Ltd

3-Fluoro-4-methoxyphenol

  • MDL number MFCD09263471
  • Cas Number : 452-11-9
  • Molecular Weight : 142.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-41.5
Synonym 2-Fluoro-4-hydroxyanisole
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-methoxyphenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-methoxyphenol
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart