APO455837596 Apollo Scientific Ltd

Silver(I) fluoride

  • MDL number MFCD00003410
  • Cas Number : 7775-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 435
Purity 98
Density 5.852
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 1150
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4705

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4705
pictograms GHS05;GHS06;GHS09
hstatements H314;H331;H400
Availability details for
Quantity
  • Availability
  • Add to Cart