APO455837597 Apollo Scientific Ltd

5-Fluoro-2-formylbenzonitrile

  • MDL number MFCD08275455
  • Cas Number : 77532-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-4-fluorobenzaldehyde
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-formylbenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-formylbenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart