APO455837602 Apollo Scientific Ltd

2-Fluoro-4-formylbenzoic acid

  • MDL number MFCD12026481
  • Cas Number : 604000-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216
Synonym 4-Carboxy-3-fluorobenzaldehyde
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45624

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45624
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart