APO455837608 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethoxy)benzonitrile

  • MDL number MFCD06660203
  • Cas Number : 886498-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid-Oil
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 60/ 5mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46663

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=46663
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart