APO455837615 Apollo Scientific Ltd

1H,1H,2'H-Perfluorodipropyl ether

  • MDL number MFCD03411203
  • Cas Number : 1000-28-8
  • Molecular Weight : 300.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.58
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 87.5
Product Link https://store.apolloscientific.co.uk/product/1h1h2h-perfluorodipropyl-ether

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h1h2h-perfluorodipropyl-ether
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart