APO455837619 Apollo Scientific Ltd

4-Bromo-2,6-difluoroiodobenzene

  • MDL number MFCD01861126
  • Cas Number : 160976-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-42
Synonym 5-Bromo-1,3-difluoro-2-iodobenzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-26-difluoroiodobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-26-difluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart