APO455837625 Apollo Scientific Ltd

3-(Bromomethyl)-2'-fluorodiphenyl ether

  • MDL number MFCD00059914
  • Cas Number : 242812-04-0
  • Molecular Weight : 281.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[3-(Bromomethyl)phenoxy]-2-fluorobenzene, 3-(2-Fluorophenoxy)benzyl bromide
Density 1.46
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromomethyl-2-fluorodiphenyl-ether

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromomethyl-2-fluorodiphenyl-ether
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart