APO455837634 Apollo Scientific Ltd

1-Fluoro-2-(trifluoromethoxy)benzene

  • MDL number MFCD00236322
  • Cas Number : 2106-18-5
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluorophenyl trifluoromethyl ether, alpha,alpha,alpha,2-Tetrafluoroanisole
Purity 98
Density 1.3
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 105-106
Product Link https://store.apolloscientific.co.uk/product/1-fluoro-2-trifluoromethoxybenzene

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-fluoro-2-trifluoromethoxybenzene
pictograms GHS02;GHS07
hstatements H225;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart