APO455837652 Apollo Scientific Ltd

6-[4-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine

  • MDL number MFCD00268940
  • Cas Number : 186834-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239-241
Synonym 2,4-Diamino-6-[(4-(trifluoromethyl)phenyl)]-1,3,5-triazine, 4-(4,6-Diamino-1,3,5-triazin-2-yl)benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8197

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8197
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart