APO455837653 Apollo Scientific Ltd

Perfluorononane-1,9-dioic acid

  • MDL number MFCD00153274
  • Cas Number : 23453-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Perfluoroazelaic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 215/60mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4345

Safety Information

signalword Danger
pcodes P280;P301+P330+P331;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4345
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart