APO455837655 Apollo Scientific Ltd

Tetraethylammonium fluoride dihydrate

  • MDL number MFCD00149991
  • Cas Number : 98330-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 98
Flash Point >101
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4757

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=4757
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart