APO455837664 Apollo Scientific Ltd

2,3,5,6-Tetrafluoroaniline

  • MDL number MFCD00007644
  • Cas Number : 700-17-4
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31-33
Purity 98
Density 1.74
Flash Point 62
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 158
Product Link https://store.apolloscientific.co.uk/product/2356-tetrafluoroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2356-tetrafluoroaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart